Anti-DYRK4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7536B
80084-110EA 800.94 USD
80084-110
Anti-DYRK4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DYRK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (450–550 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR