Anti-CDK4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7520B
80084-074EA 801.36 USD
80084-074
Anti-CDK4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal (250–350 aa)
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR