Anti-CDK10 (Cyclin-dependent kinase 10) Rabbit Polyclonal Antibody

Supplier: Abgent
AP7516A
80084-066EA 801.6 USD
80084-066
Anti-CDK10 (Cyclin-dependent kinase 10) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDK10 (Cyclin-dependent kinase 10)
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR