Anti-CDC7 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7515B
80084-064EA 763.01 USD
80084-064
Anti-CDC7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC7
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal (500–600 aa)
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR