Anti-DAPK2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7218A
80083-924EA 801.16 USD
80083-924
Anti-DAPK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal V55 (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR