Anti-PRKCG Rabbit Polyclonal Antibody

Supplier: Abgent
AP7021A
80083-808EA 801.16 USD
80083-808
Anti-PRKCG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (320–336 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR