Anti-USP14 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2142B
80083-672EA 762.8 USD
80083-672
Anti-USP14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: USP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal ( 400–475 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR