Anti-E2EPF Rabbit Polyclonal Antibody

Supplier: Abgent
AP2120B
80083-588EA 801.16 USD
80083-588
Anti-E2EPF Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: E2EPF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR