Anti-UBE2L3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2117B
80083-576EA 801.6 USD
80083-576
Anti-UBE2L3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UBE2L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal ( 100–200 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR