Anti-CDC34 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2116B
80083-572EA 763.01 USD
80083-572
Anti-CDC34 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC34
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR