Anti-ZMIZ2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1251A
80083-500EA 762.8 USD
80083-500
Anti-ZMIZ2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZMIZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR