Axygen® Capillary Sequencing PCR Tubes and Caps, Corning
Supplier: Corning
TUB-310-NC
80082-928CS 683.41 USD
80082-928
Axygen® Capillary Sequencing PCR Tubes and Caps, Corning
Tubes PCR Tubes
Capillary sequencing PCR tubes and caps are for use with the ABI Prism 310 sequencer.
  • Made of 99.9% pure, virgin polypropylene
  • Certified RNase-, DNase-, and endotoxin-free
  • Tubes are clear; caps are gray
All Axygen PCR products are specifically designed for use within PCR thermal transfer and real time PCR applications. Ultra-thin and consistent wall thickness allows for precise thermal transfer along the entire surface of the tube. The unique cap design guarantees a perfect fit to ensure there is no sample evaporation during thermal cycling.