Neptune 70-118 Kevlar® Sleeve, Ansell
Supplier: Ansell Healthcare
222146
80080-585CS 510.32 USD
80080-585
Neptune 70-118 Kevlar® Sleeve, Ansell
Workwear
Kevlar® cut- and heat-resistant sleeve.
  • Double-layered construction for extra cut resistance, with good comfort and flexibility
  • Provides forearm protection from minor heat and burn injuries
  • Thumb slot version prevents the sleeve from twisting while in use
  • Ambidextrous
Use in the automotive, glass, metal fabrication, and durable goods industries. Uses include glass handling, metal handling, and assembly.
Certifications: ANSI Level 4 Cut Protection; ANSI Level 3 Abrasion Protection.