Upward Flow Stirrer, 240 V
Supplier: Glas-Col
099DUF24
80078-732EA 2687.29 USD
80078-732
Upward Flow Stirrer, 240 V
Stirrers Overhead Stirrers
Upward flow stirrer is non-aerating, eliminating foaming and splashing during mixing.
  • 240 V model
Results for specific mixing speed ranges will vary with viscosity.
Ideal for pharmaceuticals, cosmetics, paints, inks, petroleum, and foods, this overhead stirrer provides rapid disbursement or re-suspension of heat-sensitive substances. The stirrer can also emulsify liquids of different densities, dissolve low-soluble solids, and re-suspend bacteria into solutions. Even at high speeds, the unique stirring action creates minimal surface disturbance; operation in nearly full containers of all sizes is also possible. All wetted parts are made of corrosion-resistant stainless steel for easy cleaning. A self-lubricating fluoropolymer resin bushing requires no maintenance. Shielded stirring blades protect mixing vessels from damage. The outer housing may be removed from the stirring blades for use as a conventional stirrer.
Ordering information: Complete assembly consists of a motor drive and a stirring unit. Stirrer includes a lab clamp for mounting to a lab frame.
Caution: 240 V model. Corresponding models with voltage suitable for use in US (80078-730, 80078-736) are also available, please see separate product page.