Upward Flow Stirrer, Glas-Col
Supplier: Glas-Col
099DUF12 099D25B
80078-730EA 2754.45 USD
80078-730 80078-736
Upward Flow Stirrer, Glas-Col
Stirrers Overhead Stirrers
Upward flow stirrer is non-aerating, eliminating foaming and splashing during mixing
Results for specific mixing speed ranges will vary with viscosity.
Ideal for pharmaceuticals, cosmetics, paints, inks, petroleum, and foods, this overhead stirrer provides rapid disbursement or re-suspension of heat-sensitive substances. The stirrer can also emulsify liquids of different densities, dissolve low-soluble solids, and re-suspend bacteria into solutions. Even at high speeds, the unique stirring action creates minimal surface disturbance; operation in nearly full containers of all sizes is also possible. All wetted parts are made of corrosion-resistant stainless steel for easy cleaning. A self-lubricating fluoropolymer resin bushing requires no maintenance. Shielded stirring blades protect mixing vessels from damage. The outer housing may be removed from the stirring blades for use as a conventional stirrer.
Ordering information: Complete assembly consists of a motor drive and a stirring unit. Stirrer includes a lab clamp for mounting to a lab frame.