Pulse Platform Mixer, 240 V
Supplier: Glas-Col
099A PPM24
80078-696EA 10298.8 USD
80078-696
Pulse Platform Mixer, 240 V
Shakers and Mixers Vortex Mixers
Unit accomodates a wide variety of glassware, racks, well plates, or reactor blocks and generates precise vortexing action for accurate, repeatable results.
  • 240 V model
  • Touch screen interface
The microprocessor-based control features speed display; a programmable, multifunction timer; and selectable pulse modes. The unit has chemical-resistant foam on the top and driveplate and an RS-232 communications port.
Certifications: UL listed.
Ordering information: Supplied with a 1.5 m (5'), three-wire grounded cord.
Caution: 240 V model. Corresponding model with voltage suitable for use in US (80078-694) is also available, please see separate product page.