ICON® 25 hCG Pregnancy Test, Beckman Coulter®
Supplier: BECKMAN COULTER
43025A
80078-676CS 1042.09 USD
80078-676
ICON® 25 hCG Pregnancy Test, Beckman Coulter®
Clinical Diagnostic Supplies Clinical Reagents and Kits
Highly sensitive, one-step immunochemical rapid test for qualitative detection of human chorionic gonadotropin in urine or serum to aid in early pregnancy detection.
  • Sensitive (25 mIU/mL)
  • Specificity > 99%
  • One-step (test time 3 minutes for urine, 5 minutes for serum)
  • Easy to use
Meets the requirements of physicians and hospitals for ease-of-use, value, and accuracy.
Certifications: Urine is CLIA waived; serum is moderately complex.
Ordering information: Kit contents include 25 test devices and droppers, and two additional disposable droppers.