GoldKnit® Medium-Weight Sleeve, Kevlar® Fiber, Ansell
Supplier: Ansell Healthcare
222145
80076-392CS 512.44 USD
80076-392
GoldKnit® Medium-Weight Sleeve, Kevlar® Fiber, Ansell
Workwear
SType 70-128 sleeve is knitted from 100% Kevlar® fiber.
  • Double-layered construction for extra cut resistance, with good comfort and flexibility
  • Provides forearm protection from minor heat and burn injuries
It is double-layered to provide cut-resistance, flexibility, and comfort. It also protects the forearm from minor heat and burn injuries. For convenient on-and-off, this sleeve does not have a thumb slot.