Stopcocks, High Vacuum, Double Oblique Bore, Hollow Glass Plugs, Chemglass
Supplier: Chemglass
CG-474-01 CG-474-02
80074-320EA 316.38 USD
80074-320 80074-322
Stopcocks, High Vacuum, Double Oblique Bore, Hollow Glass Plugs, Chemglass
Stopcocks
Hollow plug, double oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.