Stopcocks, High Vacuum, Single Oblique Bore, Solid Glass Plugs, Chemglass
Supplier: Chemglass
CG-473-02
80074-318EA 247.37 USD
80074-318
Stopcocks, High Vacuum, Single Oblique Bore, Solid Glass Plugs, Chemglass
Stopcocks
Solid plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug.
Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.