Stopcocks, High Vacuum, Double Oblique Bore, Glass Plugs, Chemglass
Supplier: Chemglass
CG-468-02
80074-296EA 310.1 USD
80074-296
Stopcocks, High Vacuum, Double Oblique Bore, Glass Plugs, Chemglass
Stopcocks
Solid glass high vacuum stopcock with the Chemglass constant pressure retention device.
This unique retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.
Similar to the CG-458 series, but with two parallel oblique bores. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug.