Straight Bore Stopcocks with Glass Plugs, High Vacuum, Chemglass
Supplier: Chemglass
CG-462-01 CG-462-02 CG-462-03
80074-280EA 106.56 USD
80074-280 80074-282 80074-284
Straight Bore Stopcocks with Glass Plugs, High Vacuum, Chemglass
Stopcocks
These solid glass high vacuum stopcocks feature an improved constant pressure retention device.
  • Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug
This device incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached.
Delivery information: Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kits consist of one spring and clip for the plug size.