Stopcocks, 120°, PTFE Plugs, Chemglass
Supplier: Chemglass
CG-440-01 CG-440-02
80074-224EA 187.26 USD
80074-224 80074-226
Stopcocks, 120°, PTFE Plugs, Chemglass
Stopcocks
A 1:5 taper PTFE stopcock with three sidearms sealed 120° apart. Plug has a bore to allow connection of any two arms at once.