Stopcocks, T-Bore, Capillary Arms, PTFE Plugs, Chemglass
Supplier: Chemglass
CG-438-01
80074-222EA 190.08 USD
80074-222
Stopcocks, T-Bore, Capillary Arms, PTFE Plugs, Chemglass
Stopcocks
Similar to the CG-436 series, but with all three arms being made of 2 mm I.D. capillary tubing.