Gas Dispersion Tubes, Large, Fritted, Chemglass
Supplier: Chemglass
CG-220-01
80073-714EA 127.49 USD
80073-714
Gas Dispersion Tubes, Large, Fritted, Chemglass
Gas Tubes
Large capacity gas dispersion tube having a frit 25 mm O.D.x 70 mm long
Stem is 16 mm O.D.x 200 mm above the frit. Overall length is approximately 280 mm.