Gas Dispersion Tubes, Fritted, Chemglass
Supplier: Chemglass
CG-203-01 CG-203-02 CG-203-03 CG-203-04
80073-610EA 63.48 USD
80073-610 80073-612 80073-614 80073-616
Gas Dispersion Tubes, Fritted, Chemglass
Gas Tubes
Tubes have a hollow fritted cylinder at the end of an 8 mm O.D. tube.
Cylinder dimensions: 120O.D.x20L mm (1/2x3/4"). Overall tube length: 254 mm (10").