Vacuum Trap, Chemglass
Supplier: Chemglass
CG-4530-01
80072-656EA 852.27 USD
80072-656
Vacuum Trap, Chemglass
Vacuum Traps
Trap is designed for high volume solvent removal by vacuum.
The unique design of the stopper decreases the chance of freezing by allowing vapor to pass through the holes in the side of the inner tube. The 55/50 standard taper joint connects the trap body with the stopper. Hose connections are 15 mm OD at the largest serration.