Reaction Vessel Lid, 3-Necks, Chemglass
Supplier: Chemglass
CG-1940-A-01
80071-582EA 554.12 USD
80071-582
Reaction Vessel Lid, 3-Necks, Chemglass
Lids Reaction Vessel Lids
Reaction vessel lids feature 30° side necks.
  • Three neck reaction vessel lid
  • To fit CG-1930 through CG-1931 vessels
Lid has a flat ground surface allowing it to be greased or used with an O-ring.