Powder Funnels with Offset Lower Stem, Chemglass
Supplier: Chemglass
CG-1722-03 CG-1722-01 CG-1722-04
80070-912EA 164.04 USD
80070-912 80070-908 80070-914
Powder Funnels with Offset Lower Stem, Chemglass
Funnels Powder Funnels
Funnel has a lower stem made with an offset to permit use with multi-neck flasks.