Funnels, Addition, Graduated, Glass Stopcocks, Chemglass
Supplier: Chemglass
CG-1710-01 CG-1710-03 CG-1710-06 CG-1710-15 CG-1710-02 CG-1710-04 CG-1710-10 CG-1710-13 CG-1710-16 CG-1710-19
80070-594EA 458.27 USD
80070-594 80070-598 80070-604 80070-618 80070-596 80070-600 80070-608 80070-614 80070-620 80070-626
Funnels, Addition, Graduated, Glass Stopcocks, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnels feature a double scale of numbers.
  • Graduated funnels with a double scale of numbers
  • One up from the stopcock and one down from the capacity line
With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.
Ordering information: For replacement stoppers see 10170-248, 10170-258, 80071-918, 80071-926, 80071-928, 80071-930,80071-932, 80071-934, 80071-936, 80071-938, 80071-940 and 80071-942.