Addition Funnels, Cooling Jacket, Glass Stopcock, Chemglass
Supplier: Chemglass
CG-1703-01 CG-1703-04 CG-1703-05 CG-1703-07 CG-1703-02 CG-1703-03
80070-538EA 685.01 USD
80070-538 80070-544 80070-546 80070-550 80070-540 80070-542
Addition Funnels, Cooling Jacket, Glass Stopcock, Chemglass
Funnels Dropping Funnels
Jacket permits the cooling of funnel contents by dry ice. Drain outlet has an 8 mm outer diameter. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm.