Addition Funnels, with Fluoropolymer Resin Stopcock, Chemglass
Supplier: Chemglass
CG-1702-03 CG-1702-02 CG-1702-04
80070-508EA 479.31 USD
80070-508 80070-506 80070-510
Addition Funnels, with Fluoropolymer Resin Stopcock, Chemglass
Funnels Dropping Funnels
Cylindrical style funnels feature a [ST] outer joint and lower inner joint of the same size, a pressure equalizing arm, and a [ST] fluoropolymer resin stopcock.