Volumetric Flasks with Glass Stopper, Class A, Chemglass
Supplier: Chemglass
CG-1600-01 CG-1600-02 CG-1600-05 CG-1600-06 CG-1600-09 CG-1600-10 CG-1600-03 CG-1600-04 CG-1600-07 CG-1600-08
80070-346EA 54.95 USD
80070-346 80070-348 80070-354 80070-356 80070-362 80070-364 80070-350 80070-352 80070-358 80070-360
Volumetric Flasks with Glass Stopper, Class A, Chemglass
Flasks Volumetric Flasks
Type 1, Class “A” unserialized flasks.
Certifications: Conform to Federal Specifications NNN-F-289 for Type 1, Class “A” unserialized flasks and ASTM E-288.
Delivery information: Supplied complete with one solid glass pennyhead stopper.