Rotary Evaporator Bump Traps, Modified, Chemglass
Supplier: Chemglass
CG-1322-12
80068-118EA 265.61 USD
80068-118
Rotary Evaporator Bump Traps, Modified, Chemglass
Vacuum Traps
Trap provides expanded volume for reactions that have a tendency to foam excessively.
  • Traps have a 24/40 top outer joint and the listed size of lower inner joint
  • Trap prevents entry into the condensers
The inner vapor tube is sealed to the top of the trap to prevent entry into the condenser.