Rotary Evaporator Bump Trap, Self-Washing, Chemglass
Supplier: Chemglass
CG-1319-05
80068-056EA 276.58 USD
80068-056
Rotary Evaporator Bump Trap, Self-Washing, Chemglass
Vacuum Traps
Trap is designed to eliminate hold up common to the round style of rotary evaporator traps.
  • Tapered sidewalls permit continuous washing with condensed solvent
  • While preventing vapor cooling points along the path of the condenser
The upper stem has four large holes flush with the lower end allowing solvent vapor to flow freely and making this portion of the trap easy to clean. The increased volume of the trap prevents 'bumping' and keeps solvent out of the vapor tube.