Schwartz Drying Tubes, Chemglass
Supplier: Chemglass
CG-1297-01
80068-002EA 307.46 USD
80068-002
Schwartz Drying Tubes, Chemglass
Tubes Drying Tubes
Schwartz type drying tube having hollow stoppers.
Stoppers have a hole that aligns with a serrated hose connection for on/off action.