Drying Apparatus, Abderhalden, Modified, Chemglass
Supplier: Chemglass
CG-1290-05
80067-966EA 1586.47 USD
80067-966
Drying Apparatus, Abderhalden, Modified, Chemglass
Desiccators Desiccators, Glass
Abderhalden feature a 57 mm flat O-ring connection.
  • Modified Abderhalden drying apparatus
  • Desiccant flask and a drying chamber permit easy sample removal after drying
The drying chamber has an I.D. of 2" and is 8" long. The desiccant flask has a 4 mm glass stopcock and a serrated connection for the vacuum source. The flask and condenser joints are 24/40. Complete apparatus consists of a drying chamber, desiccant flask, condenser, clamp and Viton O-ring.