Distillation Receiver, Graduated, Chemglass
Supplier: Chemglass
CG-1275-14
80067-874EA 578.71 USD
80067-874
Distillation Receiver, Graduated, Chemglass
Distillation Components Distillation Receivers
Graduated distillation receivers feature a 4 mm three way stopcock.
  • The 135° angle between the center line of the condenser
  • Receiver tubes allows the distillate to fall directly into the receiver tubes
This receiver can be used with short path style distilling heads.
Delivery information: Complete receiver is supplied with distribution adapter, 3×12 ml receiver tubes, and 6×CG-110 stainless steel springs.