Distillation Receivers, Graduated, Chemglass
Supplier: Chemglass
CG-1272-01
80067-856EA 1129.66 USD
80067-856
Distillation Receivers, Graduated, Chemglass
Distillation Components Distillation Receivers
Graduated distillation receiver with a 4 mm three way stopcock to permit solvent removal through the 10 mm outer diameter tube or return to distillation flask
Upper sampling port has a 2 mm PTFE stopcock. The top outer and lower inner joints are the same size. Solvent recovery with 24/40 joint size.