Reservoirs, Chromatography, Rodaviss® Joints, Chemglass
Supplier: Chemglass
CG-1194-01 CG-1194-05 CG-1194-06 CG-1194-07 CG-1194-11 CG-1194-02 CG-1194-03 CG-1194-04
80066-876EA 167.18 USD
80066-876 80066-884 80066-886 80066-888 80066-892 80066-878 80066-880 80066-882
Reservoirs, Chromatography, Rodaviss® Joints, Chemglass
Adapters Adapters, Joints with Tube Connectors
Reservoir for use with chromatography columns (CG-1192 and CG-1193).
  • With 24/40 lower inner joint
Reservoir has a top 24/40 standard taper outer joint for use with CG-1195 flow controller, and the listed size RODAVISS® lower inner joint.
Delivery information: Supplied complete with one CG-182 open top cap, one CG-305 Viton O-ring and one CG-184 loosening ring.