Inlet Adapters, Straight, with Hose Connection, Chemglass
Supplier: Chemglass
CG-1062-01 CG-1062-14
80066-122EA 97.49 USD
80066-122 80066-126
Inlet Adapters, Straight, with Hose Connection, Chemglass
Adapters Adapters, Joints with Tube Connectors
With serrated hose connection having an O.D. of 10 mm at the largest serration sealed between the top outer and lower inner standard taper joints.