Gas Inlet Adapter, with Hose Connection, Chemglass
Supplier: Chemglass
CG-1060-01
80066-118EA 147.39 USD
80066-118
Gas Inlet Adapter, with Hose Connection, Chemglass
Adapters Adapters, Joints with Tube Connectors
Gas inlet adapter with a tube extending 200 mm below the inner joint.
  • 24/40 joint size
  • With serrated hose connection having an O.D. of 10 mm at the largest serration
Permits gas to be introduced at the bottom of larger reaction vessels or flasks.