Thermometer Adapter, 10° Offset, Chemglass
Supplier: Chemglass
CG-1054-01
80066-102EA 93.63 USD
80066-102
Thermometer Adapter, 10° Offset, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter has a 10/30 outer joint at the top for use with thermometers.
  • Allows angling of thermometers into bottom of the flask
  • 10° offset
When used with vertical side neck flasks, thermometers can be angled into the bottom of the flask.