Flow Control Adapter, PTFE Stopcock, 90°, Chemglass
Supplier: Chemglass
CG-1030-A-01
80065-914EA 172.74 USD
80065-914
Flow Control Adapter, PTFE Stopcock, 90°, Chemglass
Adapters Adapters, Joints with Tube Connectors
Flow control adapter, used to regulate the flow of gases or liquids into reaction systems.
  • Equipped with a PTFE stopcock
  • With standard taper outer joint
Stopcock has a bore of 2 mm. Hose connection has an O.D. of 8 mm at the middle serration.