90° Flow Control Adapters with Fluoropolymer Resin Stopcock, Chemglass
Supplier: Chemglass
CG-1030-01 CG-1030-02 CG-1030-14
80065-900EA 208.05 USD
80065-900 80065-902 80065-908
90° Flow Control Adapters with Fluoropolymer Resin Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
This adapter is desgined to regulate the flow of gases or liquids into reaction systems.
  • 90° angle bend
  • Fluoropolymer resin stopcock
  • Inner [ST] joint
  • With hose connection
Stopcock has a bore of 2mm. Hose connection has an 8mm O.D. at the middle serration.