Flow Control Adapters, 90°, with Glass Stopcock, Chemglass
Supplier: Chemglass
CG-1028-01 CG-1028-02 CG-1028-19 CG-1028-29
80065-866EA 140.58 USD
80065-866 80065-868 80065-876 80065-878
Flow Control Adapters, 90°, with Glass Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Flow control adapters, used to regulate the flow of gases or liquids into reaction systems.
  • Supplied with a retaining clip
  • Equipped with 2 mm glass stopcock
Stopcock has a bore of 2 mm and is supplied complete with a retaining clip. Hose connection has an O.D. of 8 mm at the middle serration.