Schlenk In-Line Bubbler, Airfree, Chemglass
Supplier: Chemglass
AF-0510-20
80064-554EA 142.77 USD
80064-554
Schlenk In-Line Bubbler, Airfree, Chemglass
Bubblers
Used to monitor gas flow to reactions while providing a vent to the atmosphere
Bubbler has a "T" connection at the top to allow it to be mounted in-line while the reservoir head prevents oil from being sucked back into the system. Volume below the sidearm is approx 40 mL. Bubbler has 8 mm OD top and side tubulations.