Airfree® Schlenk Manometer/Bubbler, Chemglass
Supplier: Chemglass
AF-0453-02 AF-0453-04 AF-0453-05
80060-124EA 406.26 USD
80060-124 80060-126 80064-442 80064-444
Airfree® Schlenk Manometer/Bubbler, Chemglass
Bubblers
This mercury manometer/bubbler is a useful accessory for vacuum/inert systems.
  • (80060-124) has two reservoirs, while the (80060-126) has only one reservoir
  • Top has a bump trap to prevent mercury from contaminating the system
  • Hose connections have an 11 mm O.D. at the largest serration
This manometer/bubbler can be used for Schlenk manipulations, transfer of approximate quantities of gases, and for measurement of intermediate pressures. Both sides may be evacuated to ensure an oxygen-free environment. The double reservoir design allows a smaller overall volume of mercury to be used for both exhausting the excess inert gas and to fill the tube on the vacuum side.