Lattice Connectors, Chemglass
Supplier: Chemglass
CG-9260-01
80063-660EA 30.25 USD
80063-660
Lattice Connectors, Chemglass
Support Rods and Frames
Angled adjustment screws allow easy set-up and prevent slippage or misalignment.
Two U-shaped slots permit rods or clamps of 1/2” (13 mm) maximum diameter to be joined in a 90° corner joint or cross-connection with components in close proximity to minimize wasted space. Each rod or clamp adjusts independently for easy installation.