Clamp Holder, 19 mm Grip, Chemglass
Supplier: Chemglass
CG-9250-01
80063-652EA 36.56 USD
80063-652
Clamp Holder, 19 mm Grip, Chemglass
Clamps General Purpose Clamps
Nickel-plated zinc alloy clamp holder has high tensile strength and corrosion resistance.
  • Useful wherever clamping at 90° is required
  • Large thumb screws allow easy adjusting